missing translation for 'onlineSavingsMsg'
Learn More
Gold nanoparticles, 20nm, amine-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific Chemicals Product Code.: 15414645

Gold nanoparticles, 20nm, amine-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific Chemicals

Product Code. 15414645
0.5 mL
Click to view available options
Quantity:
0.5 mL
1 mL
This item is not returnable. View return policy

Product Code. 15414645

Brand: Thermo Scientific Alfa Aesar J67465.&I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Red
Quantity 0.5 mL
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, amine-functionalized, 3kDa PEGylated, OD: 50, 524nm absorption

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU22K005 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberJ67465.&I,J67465.XF
Lot NumberQ09K031 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberJ67465.&I,J67465.XF
Lot NumberZ04J012 Certificate TypeCertificate of Analysis Date28/12/2023 Catalog NumberJ67465.&I,J67465.XF
Lot NumberQ19J036 Certificate TypeCertificate of Analysis Date25/04/2023 Catalog NumberJ67465.&I
Lot NumberY19H013 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ67465.&I,J67465.XF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold nanoparticles, 20nm, amine-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.