missing translation for 'onlineSavingsMsg'
Learn More
Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption, Thermo Scientific Chemicals Product Code.: 15495835

Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption, Thermo Scientific Chemicals

Product Code. 15495835
25 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 15495835

Brand: Thermo Scientific Alfa Aesar J67269.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

Solubility
Fully miscible in water. It is soluble in aqua regia, nitric acid and hydrochloric acid.

Notes
Particles per ml: 6.54E+11
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Pink to Red
Quantity 25 mL
Solubility Information Fully miscible in water. It is soluble in aqua regia,nitric acid and hydrochloric acid.
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 524nm absorption

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP12L019 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberJ67269.AC,J67269.AE
Lot NumberN18L060 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberJ67269.AC,J67269.AE
Lot NumberM17L022 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ67269.AC,J67269.AE
Lot NumberM08L002 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog NumberJ67269.AC,J67269.AE
Lot NumberZ06K019 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberJ67269.AC,J67269.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.