missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 20nm, supplied in 0.1mM PBS, 95%, OD1, 524 nm absorption, Thermo Scientific Chemicals

Product Code. 15494585
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Red
Quantity 100 mL
Percent Purity 95%
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS, 95%, OD: 1, 524 nm absorption

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'