missing translation for 'onlineSavingsMsg'
Learn More
Gold nanoparticles, 40nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 530nm absorption, Thermo Scientific Chemicals Product Code.: 15414485

Gold nanoparticles, 40nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 530nm absorption, Thermo Scientific Chemicals

Product Code. 15414485
25 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 15414485

Brand: Thermo Scientific Alfa Aesar J67001.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Pink to Red
Quantity 25 mL
Odor Odorless
Particle Size 40nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 530nm absorption

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP12L009 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberJ67001.AC,J67001.AE
Lot NumberP12L008 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberJ67001.AC,J67001.AE
Lot NumberN03L005 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberJ67001.AC,J67001.AE
Lot NumberM08L001 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog NumberJ67001.AC,J67001.AE
Lot NumberZ09K001 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberJ67001.AC,J67001.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold nanoparticles, 40nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 530nm absorption, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.