missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 50nm, supplied in 0.1 mg/ml sodium citrate with stabilizer OD1, 535nm absorption

Product Code. p-7057851
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 15404635

Brand: Thermo Scientific Alfa Aesar J67433.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-57-5 | Au | 196.97 g/mol

For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Pink to Red
Quantity 25 mL
Particle Size 50nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer OD: 1, 535nm absorption
My Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer have turned a clear to bluish color. What causes this?

Irreversible aggregation is characterized by a clear to bluish solution upon the addition of salt. Since non-functionalized gold nanoparticles are sensitive to salt-containing buffers, re-suspension should always be performed in ultra-pure water to prevent irreversible aggregation.

How can I wash Gold nanoparticles, supplied in 0.1 mg/ml sodium citrate with stabilizer?

Although it is not generally necessary to wash the gold nanoparticles prior to use, some applications may require additional washing procedures. The easiest way to remove possible contaminants in the nanoparticles solution is by centrifugation. Centrifugation force is dependent on size of the gold nanoparticles. We recommend the following to wash gold nanoparticles:
1. Place 1 mL aliquot of colloidal gold in a 1.5 mL microcentrifuge tube.
2. Centrifuge the gold nanoparticles for 30 minutes using the recommended G force based on particle size (see below).
3. Remove the supernatant and re-suspend in an appropriate volume of ultra-pure water.
4. Vortex to re-disperse the particles.

Centrifugation speed guide for nanoparticle size:
size - G force
5 nm - 100,000 g
10 nm - 17,000 g
15 nm - 17,000 g
20 nm - 6,500 g
30 nm - 4,500 g
40 nm - 2,500 g
50 nm - 2,000 g
60 nm - 1,125 g
80 nm - 600 g
100 nm - 400 g

Do I need to wash Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer before use?

Although it is not generally necessary to wash the gold nanoparticles prior to use, some applications may require additional washing procedures. The easiest way to remove possible contaminants in the nanoparticles solution is by centrifugation. Centrifugation force is dependent on size of the gold nanoparticles. We recommend the following to wash gold nanoparticles:
1. Place 1 mL aliquot of colloidal gold in a 1.5 mL microcentrifuge tube.
2. Centrifuge the gold nanoparticles for 30 minutes using the recommended G force based on particle size (see below).
3. Remove the supernatant and re-suspend in an appropriate volume of ultra-pure water.
4. Vortex to re-disperse the particles.

Centrifugation speed guide for nanoparticle size:
size - G force
5 nm - 100,000 g
10 nm - 17,000 g
15 nm - 17,000 g
20 nm - 6,500 g
30 nm - 4,500 g
40 nm - 2,500 g
50 nm - 2,000 g
60 nm - 1,125 g
80 nm - 600 g
100 nm - 400 g

What are the recommended storage conditions for the Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer?

We recommend 4°C storage for our Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer. Do not freeze the particles because this causes irreversible aggregation. When stored for a long period of time, gold nanoparticles may sediment at the bottom of the flask, which is especially true for larger particle sizes. Prior to use, re-suspend the sedimented particles by swirling until a homogenous solution is obtained.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.