Learn More
Gold nanoparticles, 50nm, supplied in 0.1mM PBS, 95%, OD1, 535nm absorption, Thermo Scientific Chemicals
Description
For conjugate development by passive adsorption of proteins and other ligands to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For conjugate development by passive adsorption of proteins and other ligands to the gold nanoparticle surface
- Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

Specifications
Specifications
Color | Pink to Red |
Quantity | 25 mL |
Percent Purity | 95% |
Particle Size | 50nm |
Sensitivity | Light sensitive |
Form | Liquid |
Chemical Name or Material | Gold nanoparticles, supplied in 0.1mM PBS, 95%, OD: 1, 535nm absorption |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N18L055 | Certificate of Analysis | 20/02/2025 | J67157.AC,J67157.AE |
Z06K024 | Certificate of Analysis | 11/12/2024 | J67157.AC,J67157.AE |
Z06K020 | Certificate of Analysis | 11/12/2024 | J67157.AC,J67157.AE |
X29K031 | Certificate of Analysis | 13/11/2024 | J67157.AC,J67157.AE |
W10K026 | Certificate of Analysis | 23/09/2024 | J67157.AC,J67157.AE |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.