missing translation for 'onlineSavingsMsg'
Learn More
Gold nanoparticles, 5nm, supplied in 0.1mM PBS, 95%, OD1, 515-520nm absorption, Thermo Scientific Chemicals Product Code.: 15454515

Gold nanoparticles, 5nm, supplied in 0.1mM PBS, 95%, OD1, 515-520nm absorption, Thermo Scientific Chemicals

Product Code. 15454515
25 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 15454515

Brand: Thermo Scientific Alfa Aesar J67089.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold sputtering target metal is retained for use as bullion reserves, but some is used within the electronics and jewellery industries, where it is frequently alloyed with other elements to improve the mechanical properties of the metal (e.g. copper and silver). Other uses for the metal are as a heat reflecting coating for glass as well as as a decorative medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gold sputtering target metal is retained for use as bullion reserves, but some is used within the electronics and jewellery industries, where it is frequently alloyed with other elements to improve the mechanical properties of the metal (e.g. copper and silver). Other uses for the metal are as a heat reflecting coating for glass as well as as a decorative medium.

Solubility
Insoluble in water. It is soluble in aqua regia, Hno3/Hcl.

Notes
Size dispersity <15%. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Thickness tolerance: ±0.015in; Diameter tolerance: +0.020/-0.0in.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Red
Quantity 25 mL
Solubility Information Insoluble in water. It is soluble in aqua regia,Hno3/Hcl.
Percent Purity 95%
Odor Odorless
Particle Size 5nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS, 95%, OD: 1, 515 to 520nm absorption

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP12L025 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberJ67089.AE,J67089.AC
Lot NumberP05L025 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog NumberJ67089.AE,J67089.AC
Lot NumberN18L056 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberJ67089.AE,J67089.AC
Lot NumberW16K015 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog NumberJ67089.AE,J67089.AC
Lot NumberU09K034 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog NumberJ67089.AE,J67089.AC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold nanoparticles, 5nm, supplied in 0.1mM PBS, 95%, OD1, 515-520nm absorption, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.