missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 5nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 515-520nm absorption, Thermo Scientific Chemicals

£86.73 - £253.26

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15474635
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J67448.AC
25 mL
£86.73
25mL
Please to purchase this item. Need a web account? Register with us today!
15484635
View Documents
Thermo Scientific Alfa Aesar
J67448.AE
100 mL
£253.26
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

Red
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
Odorless
Light sensitive
Gold nanoparticles, supplied in 0.1mM PBS, reactant to free, 99%, OD1, 515 to 520nm absorption
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
99%
5nm
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only