missing translation for 'onlineSavingsMsg'
Learn More
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis) Product Code.: 15457095

Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)

Product Code. 15457095
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15457095

Brand: Thermo Scientific Alfa Aesar 045822.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-57-5 | Au | 196.97 g/mol

Gold nanopowder is intercalated within a polymer matrix lower the effective conjugation length of the polymer, resulting in a blue shift in the adsorption. It also finds application in plasmonics, electronics and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Golden
CAS 7440-57-5
Molecular Formula Au
MDL Number MFCD00003436
Quantity 1 g
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
Solubility Information Insoluble in water.
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES [Au]
IUPAC Name gold
Molecular Weight (g/mol) 196.97
PubChem CID 23985
ChEBI CHEBI:30050
Percent Purity ≥99.95%
Odor Odorless
Particle Size APS 20 to 25nm
Form Nanopowder
Assay (metals basis)
Chemical Name or Material Gold nanopowder
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH20Y038 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number045822.03
Lot NumberJ28X033 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045822.03
Lot NumberH20Y038 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045822.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.