missing translation for 'onlineSavingsMsg'
Learn More

Gold Palladium wire, 0.2mm (0.008in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

£71.25 - £270.35

Specifications

Chemical Name or Material Gold Palladium wire
Name Note 0.2mm (0.008 in.) dia., Annealed, Temper: softest 0.154g/ft
Avg. Mol. Wt. or Mol. Wt. Range Au:Pd; 60:40 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD00003436
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349577
View Documents
Thermo Scientific Alfa Aesar
041206.BU
25 cm
£71.25
25cm
Please to purchase this item. Need a web account? Register with us today!
11339577
View Documents
Thermo Scientific Alfa Aesar
041206.BW
50 cm
£153.77
50cm
Please to purchase this item. Need a web account? Register with us today!
11329577
View Documents
Thermo Scientific Alfa Aesar
041206.G1
1 m
£270.35
1m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gold Palladium wire is used as a microelectrode device, which exhibits excellent responses to hydrogen in concentrations as low as 0.5% by volume. It is also involved in the enhanced sensitivity for the detection of hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold Palladium wire
Au:Pd; 60:40 wt%
MFCD00003436
99.9%
0.2mm (0.008 in.) dia., Annealed, Temper: softest 0.154g/ft
(metals basis)
Insoluble in water.
Wire
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold Palladium wire, 0.2mm (0.008in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.