missing translation for 'onlineSavingsMsg'
Learn More

Gold plating solution, electroless, metal content≈3.7g/L, Thermo Scientific Chemicals

For coating nickel or copper

£251.90 - £947.72

Specifications

Chemical Name or Material Gold plating solution
Color Colorless to Yellow
Name Note Electroless, metal content ≈3.7g/L
MDL Number MFCD00003436
UN Number UN2922
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324358
View Documents
Thermo Scientific Alfa Aesar
042307.AK
250 mL
£251.90
250mL
Please to purchase this item. Need a web account? Register with us today!
11334358
View Documents
Thermo Scientific Alfa Aesar
042307.K2
1 L
£947.72
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For coating nickel or copper

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold plating solution
Electroless, metal content ≈3.7g/L
UN2922
Colorless to Yellow
MFCD00003436
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H311-H314-H332-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold plating solution, electroless, metal content≈3.7g/L, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.