missing translation for 'onlineSavingsMsg'
Learn More
Gold powder, spherical, APS 1.5-3.0 micron, 99.96+% (metals basis), Thermo Scientific Chemicals Product Code.: 11379127

Gold powder, spherical, APS 1.5-3.0 micron, 99.96+% (metals basis), Thermo Scientific Chemicals

Product Code. 11379127
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11379127

Brand: Thermo Scientific Alfa Aesar 039818.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Apparent density 40-70g/in3, Tap density: 4.0-9.0g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Assay Percent Range 99.96+%, (metal basis)
Quantity 5 g
Form Spherical Powder
Chemical Name or Material Gold

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW10K031 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog Number039818.MD,039818.06,039818.03
Lot NumberT09K010 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number039818.MD,039818.06,039818.03
Lot NumberZ14I006 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number039818.MD,039818.06,039818.03,039819.04
Lot NumberU25H008 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number039818.MD,039818.06,039818.03,039819.04
Lot NumberN23I067 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039818.MD,039818.06,039818.03,039819.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold powder, spherical, APS 1.5-3.0 micron, 99.96+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.