missing translation for 'onlineSavingsMsg'
Learn More

Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196001000


 View more versions of this product

Product Code. 10666212

  • £40.30 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gold standard solution
1 mg/ml Au in 0.5N HCl
Plastic bottle
Orange/Yellow to Yellow
MFCD00003436
[Au]
196.97
CHEBI:30050
AAS
1.09
Powder and/or Chunks
990 to 1010μg/mL of Au(3+) in 10 to 20% HCl
7440-57-5, 7647-01-0, 7732-18-5
100 mL
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
196.97
1.0900g/mL
Soluble in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gold standard solution, 1 mg/ml Au in 10-20% HCl

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only