missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Blank Standard 1 For ICP/MS, SPEX CertiPrep™
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Au | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 7440-57-5, 7647-01-0, 7732-18-5 |
Packaging | Triple-leached LDPE bottle |
CAS Min % | 0.01, 2.0, 97.99 |
Quantity | 125 mL |
CAS Max % | 0.01, 2.0, 97.99 |
Certificate of Analysis | Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Molecular Formula | Au |
MDL Number | MFCD00003436 |
Show More |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'