missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 0.25mm (0.01in) dia, 99.9% (metals basis)

Product Code. 11386627
Click to view available options
Quantity:
1 m
5 m
Unit Size:
1m
5m
This item is not returnable. View return policy

Product Code. 11386627

Brand: Thermo Scientific Alfa Aesar 000725.G5

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold wire
Name Note 0.25mm (0.01 in.) dia.
CAS 7440-57-5
Quantity 5 m
Odor Odorless
Weight ≈0.95g/m
Assay Percent Range 99.9% (metals basis)
Assay Percent Notes (metals basis)
Molecular Formula Au
MDL Number MFCD00003436
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES [Au]
IUPAC Name gold
Molecular Weight (g/mol) 196.97
PubChem CID 23985
ChEBI CHEBI:30050
Percent Purity 99.9%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.