missing translation for 'onlineSavingsMsg'
Learn More
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11307048

Gold wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11307048
50 cm
Click to view available options
Quantity:
2 cm
10 cm
50 cm
This item is not returnable. View return policy

Product Code. 11307048

Brand: Thermo Scientific Alfa Aesar 014726.BW

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.15.15g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored wire
Assay Percent Range 99.999% (Metals basis)
Quantity 50 cm
Odor Odorless
Form Wire
Chemical Name or Material Gold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.