missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Autoclave Bags Printed with Biohazard Logo
Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.
Brand: Corning™ SA03-10
22.14 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
- Polypropylene (PP) resistant bags
- Thermal resistance 134°C
- Transparent
Specifications
3 L | |
40 μm | |
1/Pk. | |
400 mm |
Polypropylene | |
500/Cs. | |
255 mm |