Learn More
Corning™ Autoclave Bags Printed with Biohazard Logo
Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.
Marke: Corning™ SA09-10
22.78 GBP valid until 2024-12-31
Use promo code "23288" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00350kg
Beschreibung
- Polypropylene (PP) resistant bags
- Thermal resistance 134°C
- Transparent
Spezifikation
9 L | |
40 μm | |
1/Pk. | |
500 mm |
Polypropylene | |
500/Cs. | |
300 mm |