missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Autoclave Bags Printed with Biohazard Logo
Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.
Brand: Corning™ SA21-10
52.29 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polypropylene (PP) resistant bags
- Thermal resistance 134°C
- Transparent
Specifications
21 L | |
40 μm | |
1/Pk. | |
780 mm |
Polypropylene | |
500/Cs. | |
400 mm |