Learn More
Gosselin™ Inoculating L-Shaped Spreaders
GREENER_CHOICE
For spreading over the surface of solid media in microbiological applications. Gosselin™ Inoculating L-Shaped Spreaders are manufactured from polystyrene.
Brand:  Gosselin™ ETAR-01
79.03 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 12908140
  • £156.00 / Pack of 1000
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Curved end
  • Sterile and disposable
  • Packed in tamper-evident ZIP bags
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
White
Pack of 1000
Polystyrene
Sterile
EC N°1935/2004, ISO 11137
148 mm
L-Shape
Inoculating Spreader
For Dairy, Environmental, Industrial, Microbiological Uses
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gosselin™ Inoculating L-Shaped Spreaders > 150 bags of 20