Learn More
Gosselin™ Conical Containers
Provides safe and secure storage in the laboratory. Gosselin™ Conical Containers are manufactured from either clear or white polypropylene.
Brand:  Gosselin™ PC200A-05
 View more versions of this product
Product Code. 11819002
  • £128.30 / Pack of 660
Explore more special offers
Description
Description
  • Shock resistant
  • Molded graduations outside on clear containers; molded internal graduations on white containers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Container
Yes
Conical, Cylindrical
200 mL
Natural
25 mL
660 /Cs.
No
Polypropylene
Non-sterile
CE, IVD, EC N°1935/2004
62 mm
88 mm