missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Straw Pipets, Polypropylene, Sterile
GREENER_CHOICE

Decrease cross contamination risks using disposable pipets

Brand:  Corning™ EPI1B-05

 View more versions of this product

Product Code. 11962785

  • £375.04 / Pack of 12000
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene straw pipet
  • Packed in tamper evident ZIP bags
  • For liquid sampling
  • Universal size
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sterility: SAL 10-3
Sterile
100/Bag
19 cm
Single-use
Polypropylene
100/Bag
1 mL
Straw Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.