Learn More
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
£2580.93 - £4554.94
Specifications
Display Type | VFD |
---|---|
Stabilization Time | 1.5 seconds |
Type | Industrial Precision Balance |
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
16601132
|
A&D Weighing
GP-30K-EC |
31 kg | 0.1 g |
£3783.01
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16621132
|
A&D Weighing
GP-32K-EC |
31 kg/6.1 kg | 1 g/0.1 g (smart range) |
£2999.30
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16651122
|
A&D Weighing
GP-100K-EC |
101 kg | 1 g |
£4554.94
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16651132
|
A&D Weighing
GP-60K-EC |
61 kg | 1 g |
£3783.01
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16671122
|
A&D Weighing
GP-102K-EC |
101 kg/61 kg | 10 g/1 g (smart range) |
£4048.17
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16681122
|
A&D Weighing
GP-12K-EC |
12 kg | 0.1 g |
£2580.93
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16691122
|
A&D Weighing
GP-20K-EC |
21 kg | 0.1 g |
£2851.99
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- EC Type approved
- Swing arm design or separate display and platform design
- IP65 protected
- Durable aluminum alloy & stainless steel construction
- Easy calibration with internal calibration mass
- SHS response speed of only 1.5 seconds
- GLP, GMP and ISO compliant
- RS-232C interface
- Comparator output and analog output (optional)
- Large Vacuum Fluorescent display
- Data memory function
- Counting function with ACAI (Automatic Counting Accuracy Improvement)
- Smart range function (GP-32K/102K/32KS)
- Comparator function
- Underhook weighing for thermal and magnetic samples as well as for density calculation
- Animal weighing function
Specifications
VFD | |
1.5 seconds | |
Industrial Precision Balance |