Learn More
A&D Weighing GP Series Precision Balances Internal Calibration (detached display)
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
Brand:  A&D Weighing GP-61KS
Additional Details : Weight : 20.00000kg
 View more versions of this product
Product Code. 16681132
  • £5226.68 / Each
Estimated Shipment: 12-08-2024
to see stock.
Description
Description
  • Swing arm design or separate display and platform design
  • IP65 protected
  • Durable aluminum alloy & stainless steel construction
  • Easy calibration with internal calibration mass
  • SHS response speed of only 1.5 seconds
  • GLP, GMP and ISO compliant
  • RS-232C interface
  • Comparator output and analog output (optional)
  • Large Vacuum Fluorescent display
  • Data memory function
  • Counting function with ACAI (Automatic Counting Accuracy Improvement)
  • Comparator function
  • Underhook weighing for thermal and magnetic samples as well as for density calculation
  • Animal weighing function
Specifications
Specifications
61 kg
0.1 g
Industrial Precision Balance
VFD
1.5 seconds