Learn More
Invitrogen™ GP2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA542593
Product Code. 15525572
  • £443.00 / 100µL
Description
Peptide sequence: SLQAALQPIV SSLNVSVDGN GEFIVRMALF QDQNYTNPYE GDAVELSVES Sequence homology: Cow: 79%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 79%; Rat: 92%.
GP2 is component of pancreatic secretory (zymogen) granule. The exact function of GP2 remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GP2
Polyclonal
Unconjugated
GP2
2310037I18Rik; AV060639; glycoprotein 2; glycoprotein 2 (zymogen granule membrane); glycoprotein 80; GP2; gp80; Pancreatic secretory granule membrane major glycoprotein GP2; pancreatic zymogen granule membrane associated protein GP2; pancreatic zymogen granule membrane protein GP-2; secretory (zymogen) granule membrane glycoprotein GP2; ZAP75; zymogen granule membrane glycoprotein 2
Rabbit
Affinity Chromatography
RUO
2813
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P55259
GP2
Synthetic peptide directed towards the middle region of human GP2.