Learn More
GPR34 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111838
Additional Details : Weight : 0.01000kg
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: CFHYRDRHNA KGE Homology: Human, rat - 12/13 amino acid residues identical
GPR34 (extracellular) Polyclonal antibody specifically detects GPR34 (extracellular) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
GPR34 (extracellular) | |
Polyclonal | |
Unconjugated | |
Gpr34 | |
G protein-coupled receptor 34; GPR34; G-protein-coupled receptor; Lypsr1; Probable G-protein coupled receptor 34 | |
Peptide corresponding to amino acid residues 197 - 209 of mouse GPR34 (2nd extracellular loop) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q9R1K6, Q9UPC5 | |
Rabbit | |
Affinity Purified | |
RUO | |
23890, 2857, 554353 | |
-20°C | |
Lyophilized |