Learn More
GPR37 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111837
Additional Details : Weight : 0.01000kg
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)RVRLKNP FYPLTQE Homology: Mouse, human amino acid residues are identical.
Parkinson is the second most common neurodegenerative disease after Alzheimers. About 1 percent of people over the age of 65 and 3 percent of people over the age of 75 are affected by the disease. The mutation is the most common cause of Parkinson disease identified to date. The function of Park2 is not well-known; however, it may play a role in the ubiquitin-mediated proteolytic pathway. Mutations in this gene are known to cause autosomal recessive juvenile parkinsonism. Alternative splicing of this gene produces three known products of undetermined function. Panneuronal expression of Parkin substrate Pael-R causes age-dependent selective degeneration of Drosophila dopaminergic (DA) neurons; coexpression of Parkin degrades Pael-R and suppresses its toxicity.Specifications
GPR37 (extracellular) | |
Polyclonal | |
Unconjugated | |
GPR37 | |
AI848630; Ednrbl; Endothelin B receptor-like protein 1; ETBR-LP-1; Gpcr/cns1; Gpr37; G-protein coupled receptor 37; G-protein coupled receptor CNS1; hET(B)R-LP; Het-b-r-lp; Pael receptor; PAELR; Pael-R; Parkin-associated endothelin receptor-like receptor; Prosaposin receptor GPR37 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117549, 14763, 2861 | |
-20°C | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O15354, Q9QY42, Q9QYC6 | |
GPR37 | |
Peptide corresponding to amino acid residues 234-247 of rat GPR37 (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |