missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR50 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | GPR50 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16859244
|
Proteintech
19762-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16849244
|
Proteintech
19762-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.Specifications
GPR50 | |
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
O88495, Q13585 | |
14765, 9248 | |
Peptide | |
Primary | |
-20°C | |
GPR50 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G protein coupled receptor 50, GPR50, H9, Mel1c, Melatonin related receptor | |
GPR50 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title