missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR62 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533745
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Marmoset (88%) Elephant, Horse (81%).
GPR62 is a protein coding gene.Specifications
GPR62 | |
Polyclonal | |
Unconjugated | |
GPR62 | |
G protein-coupled receptor 62; GPCR8; GPR62; G-protein coupled receptor 62; G-protein coupled receptor GPCR8; G-protein coupled receptor KPG_005; hGPCR8; KPG_005; MGC26943; probable G-protein coupled receptor 62 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9BZJ7 | |
GPR62 | |
Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR62. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |