Learn More
Grace™ Reveleris™ SRC C18 Cartridges
Ideal for speed, resolution and capacity
Brand:  Grace™ 145152105
 View more versions of this product
Product Code. 11353581
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Withstand higher pressures (up to 200psig) and deliver greater loading capacity
  • Superior, high load silica for excellent resolution
  • Optimally packed to give high plate count
  • Tighter particle size distribution and minimal fines keep backpressure low
  • RFID tag streamlines Reveleris system (sold separately) operation by auto-loading cartridge specifications
  • Compatible with other popular flash systems
Specifications
Specifications
C18
80 g
200 psi
Bonded Flash Cartridge
For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems