missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 100/N Smooth Surface Qualitative Filter Papers

Used for routine analyses like clarification and determination of substances

Brand:  Sartorius™ FT-2-328-580580

Product Code. 11755116

  • £104.00 / Pack of 100
Estimated Shipment: 14-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N <0.1%)
  • Wet-strengthened
  • Supplied in sheets as well as customer-specific cuts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sheet
30μm/sec.
100/N
0.18 mm
580 L x 580 Wmm
580 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100/Pk.
Qualitative Filter Paper
580 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers