Learn More
Fisherbrand™ Grade 122 General Purpose Filter Papers
A general purpose filter paper used for all general works in the laboratory for excellent and consistent filtration.
Brand:  Fisherbrand™
Code : A0
Additional Details : Weight : 0.16000kg
 View more versions of this product
Product Code. 11778742
  • £26.60 / Pack of 100
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
  • Very fast filtration velocity
  • Available in a range of sizes
  • Available as plain or folded discs
  • Applications include hospitals, research laboratories, schools, chemical industries, pharmaceuticals and more
Specifications
Specifications
Folded Disc
General works
150 mm
Extremely fast
100/Pk.