missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Grade 25 Sapphire Spheres, Dia.: 5mm
£72.30 - £991.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11365999
|
Thermo Scientific Chemicals
042058.KK |
10 Pieces |
£281.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11355999
|
Thermo Scientific Chemicals
042058.KH |
2 Pieces |
£72.30
Pack of 2 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
11379717
|
Thermo Scientific™
042058.KD |
50 Pieces |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sapphire Sphere | |
5 mm | |
Grade 25 |
RUO – Research Use Only