missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Grade 4b, Smooth Paper for Qualitative and Technical Analyses, Sheets
Sartorius Stedim Biotech™ Grade 4b, Smooth Paper for Qualitative and Technical Analyses, Sheets are suitable for analytical and technical purposes and feature medium fast filtering.
Brand: Sartorius Stedim Biotech™ FT-2-309-450450Q
This item is not returnable.
View return policy
Description
- Made of cotton linters and refined pulp with > 95% alpha-cellulose
- Medium fast filtering
- Non-sterile and bulk packed
Specifications
75 g/m2 | |
Cotton Linters, Refined Pulp | |
22 sec. | |
500/Pk. | |
Qualitative Filter Paper |
Sheet | |
8 to 12 μm | |
4 b | |
0.15 μm | |
450 x 450 mm |