missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 600 Cellulose General Purpose Filter Paper: Discs

Used in schools and as bench protective paper. Fisherbrand™ Grade 600 Cellulose General Purpose Filter Paper is economical.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.01800kg

 View more versions of this product

Product Code. 11728772

  • £5.60 / Pack of 100
Estimated Shipment: 21-08-2024
to see stock.
Add to basket

Description

Description

Applications:
  • Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifications

Specifications

Disc
10 to 20 μm
Economical filter Paper used in schools and as bench protective Paper
100/Pk.
47 mm
Cellulose
10 to 20 μm
600
0.15 mm
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers