missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 604 Surface Protection Paper

Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.

Brand:  Fisherbrand™ 0604R00023

Product Code. 11714916

  • £480.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Available in sheet or roll form
  • Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface
Specifications

Specifications

Surface Protection Paper
50 m
Polyethylene
0.25mm
Roll
Grade 604 Paper
0.92 m
White
1 roll
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers