Learn More
Fisherbrand™ Grade 604 Surface Protection Paper
Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.
Brand:  Fisherbrand™ 0604F00022
 View more versions of this product
Product Code. 11728742
  • £94.15 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Available in sheet or roll form
  • Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface
Specifications
Specifications
Surface Protection Paper
570 mm
570 x 460 mm
White
100 sheets
Grade 604 Paper
460 mm
Polyethylene
0.25mm
Sheet