Learn More
Fisherbrand™ Grade 911 Universal pH Indicator Paper
Provides rapid and precise readings for measuring pH.
Brand:  Fisherbrand™ 0911R00024
15.39 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Product Code. 11773554
  • £22.55 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Filter paper soaked with one or several colored indicators of pH
  • Dip indicator paper into the liquid to be tested for a few seconds and remove it from the test solution and compare the resultant colors with the color scale printed on the cover
  • Provided on a dispenser roll of 5m
  • pH range of 1 to 11
Specifications
Specifications
1 to 11
Measuring pH
5 m Dispenser Roll
Indicator strips