missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 912 Universal Test Paper

Used for general analysis.

Brand:  Fisherbrand™ 0912S00522

 View more versions of this product

Product Code. 11562413

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • General control of acid or alkaline reactions

Color:

  • Litmus Blue: Blue to red
  • Litmus neutral: Purple to blue/red
  • Litmus red: Red to blue

Applications:

  • Rapid determination of pH value in industrial applications
  • General analysis in school
  • Used in formation
Specifications

Specifications

Purple
Rapid determination of pH value in industrial applications, general analysis in school, formation
Neutral Litmus Paper
Tube
200 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Grade 912 Universal Test Paper > Litmus paper, neutral; Tube; Dimensions: 60 x 10mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.