missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade C 250 Refined Pulp Membrane Filter Boards
Brand: Sartorius™ FT-2-344-580580
Code : NEW
Additional Details : Weight : 0.01000kg
Spezifikation
Sheet | |
Non-sterile | |
100 Pack | |
Membrane Filter Boards |
Refined Pulp | |
Used for filtration of cooking and transformer oils, as absorptives of human serum and as base paper for further impregnation with certain reagents | |
0.43 mm | |
580 x 580mm |