missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Gradient Mixer
Ideal for pouring polyacrylamide gradient gels
Brand: Fisherbrand™ GM50
This item is not returnable.
View return policy
Description
- Pours polyacrylamide gels using sucrose or caesium chloride solution
- Acrylic plastic construction
- Leak proof valves that control mixing between reservoir and mixing chambers
- Dimensions (D x W x H): 2.4 x 9 x 9cm
- Internal dimensions of mixing and reservoir chambers (D x W): 1.3 x 8cm
- Internal diameter of outlet port: 2mm
Specifications
Pours polyacrylamide gradient gels | |
1 Each | |
2.4 x 10 x 11 cm | |
11.5 cm |
Gradient Mixer | |
50 mL | |
10 cm | |
2.4 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction