Learn More
Lenz Laborglasinstrumente™ Graduated Cylindrical Separating Funnels with PTFE Plug
Made of borosilicate glass. Lenz Laborglasinstrumente™ Graduated Cylindrical Separating Funnels with PTFE Plug are excellent for liquid-liquid extractions and separating the components of a mixture into two immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04026149
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10424291
  • £132.00 / Pack of 2
Estimated Shipment: 29-10-2024
to see stock.
Add to basket
Description
Description
  • With NS socket and PE stopper
  • With PTFE plug
  • With screwthread retaining device
  • Borosilicate glass
Specifications
Specifications
250 mL
Yes
4
Borosilicate Glass
10
2