missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Graffiti Remover 3000
Formulated to quickly and effectively remove graffiti from brickwork and concrete surfaces. Effective on a wide range of graffiti substances including aerosol spray paint, permanent markers, crayon, and emulsion paint.
Brand: 3M™ 7100030785
171.95 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Graffiti remover for plastic, painted, or varnished surfaces
- Effective against aerosol spray paint, permanent markers, lipstick, crayon, and emulsion paint
- The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
- Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
- Has minimal odor to enhance user comfort and efficiency unlike traditional chemical removers which emit powerful odors and harmful vapors
- Low environmental impact since the product does not contain aromatic, aliphatic, or chlorinated solvents
Specifications
Graffiti Remover | |
500 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 3M Graffiti Remover 3000 (New formulation)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Fatty acids, C16-18,Ethyl 3-ethoxypropionate, Dimethyl Adipate, Dimethyl Succinate
- UNKNOWN ACUTE-4 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 10% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction