Learn More
BD Gram Crystal Violet
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
£176.00 - £191.00
Specifications
Type | Crystal Violet |
---|---|
Certifications/Compliance | CE |
Format | Liquid |
For Use With (Application) | Used to stain microorganisms from cultures or specimens by the differential gram method |
Color | Purple |
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
11778193
|
BD
212525 |
Crystal Violet | 4 x 250mL |
£191.00
1 L |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11788193
|
BD
212526 |
Crystal Violet | 1 Gallon (3.8L) |
£176.00
3.80 L |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||
Description
Gram Stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.
- Complete stain kits reduce the mess associated with Gram staining procedures and include a compact tray for holding the 250mL reagent bottles
- Break-resistant bottles features no-drip, keep-clean cap spouts that make regulating the flow easy and that help to prevent spots and spills
- Color-coded labels allow quick identification of the reagent
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles
Specifications
Crystal Violet | |
Liquid | |
Purple | |
Crystal Violet |
CE | |
Used to stain microorganisms from cultures or specimens by the differential gram method | |
15° to 30°C |
Safety and Handling
- Warning
Hazard Category
- Carcinogenicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P271-Use only outdoors or in a well-ventilated area.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropanol, methanol, ethanol, C.I. basic violet 3
Your input is important to us. Please complete this form to provide feedback related to the content on this product.