missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Gram's Iodine Solution

Product Code. 12956287
Click to view available options
Quantity:
500mL
1L
2L
Unit Size:
1 L
2 L
500mL
This item is not returnable. View return policy

Product Code. 12956287

missing translation for 'mfr': Pro-Lab Diagnostics™ PL.7004

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Gram's Iodine Solution concentrates are 10 times concentrated to give a ready to use stain on dilution.

  • Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
  • Solutions come in a selection of convenient and economical sizes. 
  • All stains are manufactured in accordance with all current regulatory requirements
  • CE labelled

Specifications

Solution Type Reagent
Type Gram Iodine
Quantity 1L
Certifications/Compliance (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
Format Liquid
For Use With (Application) For Gram staining
Color Dark brown
Composition Ethanol
Stain Type Gram Stain
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.