missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Gram Staining Kit

Product Code. 12333378
Click to view available options
Includes:
Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Dilute Carbol Fuchsin 250mL
Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Neutral Red 250mL
Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Safranin 250mL
Unit Size:
0.5 L
1 L
1 set
This item is not returnable. View return policy

Product Code. 12333378

Brand: Pro-Lab Diagnostics™ PL.8055/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Gram Staining Kit contains a collection of common stains.

  • Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
  • Solutions come in a selection of convenient and economical sizes. 
  • All stains are manufactured in accordance with all current regulatory requirements
  • CE labelled

Specifications

Type Gram Stain Kit
Certifications/Compliance CE marked (98/78/EC)
Includes Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Safranin 250mL
Format Liquid
For Use With (Application) Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria
Stain Type Gram
Volume (Metric) 250 x 4 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Gram Staining Kit/Crystal Violet/Gram's Iodine/Gram's Differentiator/Safranin
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P233-Keep container tightly closed.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, C.I. basicviolet 3, methanol,ethanol, acetone, ethanol, methanol, ethanol, methanol, glyc
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.