missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Grant Bio™ Adapter Sets for R-12-10 Interchangeable Centrifuge Rotor

Accommodates a variety of vacutainers for the R-12-10 Interchangabl;e Centrifuge Rotor

Brand:  Grant Instruments™ BN-16-100

 View more versions of this product

Product Code. 15153706

  • £219.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Interchangeable centrifuge rotor for 12 x 10 to 15mL tubes
  • Rounded ends, no caps
  • Dimensions: 16mm dia. x 105mm
Specifications

Specifications

8 to 9 mL
100 mm
12/Pk.
16 mm
R-12-10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers