Learn More
Grant Instruments™ Grant Bio™ Non-Slip Rubber Mat Flat Platform
For use with ES 80 Orbital Shaking Platform
Brand:  Grant Instruments™ PP-400
Product Code. 15143603
  • £211.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with ES-80 Shaker Incubator
  • Black rubber pads provide nonslip surface for vessels on the platform.
Specifications
Specifications
Flat Platform
400 x 360 mm
ES-80