Learn More
Grant Instruments™ Grant Plastic Bath Tank
Withstands temperatures up to 99°C.
ÂŁ210.00 - ÂŁ249.10
Products 3
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Price Quantity & Availability  
11486437
View accessory products View Documents
Grant Instruments™
P12
12 L 415 x 350 x 380 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11496437
View accessory products View Documents
Grant Instruments™
P18
18 L 600 x 350 x 380 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11476437
View accessory products View Documents
Grant Instruments™
P5
5 L 390 x 200 x 380 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Withstands temperatures up to 99°C.
  • Can be combined for use with any Grant digital thermostat or heated circulating bath
  • Plastic construction
  • 85/140mm min/max liquid depth
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Grant Instruments™ Grant Plastic Bath Tank