missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Granulate Universal Fine Grain

Granulate Universal Fine Grain is ideal for setting oils, alkalis, lipids, fuels, lacquers etc.

Brand:  asecos™ 9906

Additional Details : Weight : 20.00000kg

 View more versions of this product

Product Code. 17886526

  • £87.45 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket

Description

Description

  • 0.5-1 mm grain size
  • 20 kg bag binds up to 18 L heating oil
  • Penetrates small depressions and cracks
  • Best suited for large contact with the surface and fast absorption (e.g. traffic surfaces)
  • Optimum slip resistance even when completely saturated
  • Available in one or three 20 kg bags.
Specifications

Specifications

Loose Particulate
Absorbent Granules
44.09 lb.
20 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers